2-(Trifluoromethoxy)benzoic acid ≥97%

Supplier: Matrix Scientific

005311-5G 005311-25G
101788-656EA 20 USD
101788-656 101788-658
2-(Trifluoromethoxy)benzoic acid ≥97%
2-(Trifluoromethoxy)benzoic acid

Hazard: Irritant

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD00052325
CAS Number: 1979-29-9

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR