tert-Butyl-4-(5,6-dichloropyridine-3-amido)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-{[(5,6-dichloropyridin-3-yl)carbonyl]amino}piperidine-1-carboxylate
059825-500MG
059825-1G
059825-5G
101919-220EA
600
USD
101919-220
101919-222
101919-224
tert-Butyl-4-(5,6-dichloropyridine-3-amido)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(5,6-dichloropyridine-3-amido)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁Cl₂N₃O₃ Melting Pt: 201…202 °C |
MDL Number:
MFCD18904320 CAS Number: 1296172-35-4 |
Specification Test Results
Melting point | 201-202°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...