3-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}-3-azabicyclo-[3.1.0]hexane-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

059824-500MG 059824-1G
101919-216EA 1396.92 USD
101919-216 101919-218
3-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}-3-azabicyclo-[3.1.0]hexane-2-carboxylic acid ≥95%
3-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}-3-azabicyclo-[3.1.0]hexane-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₄O₂
Melting Pt: 183…184 °C
MDL Number: MFCD18904314
CAS Number: 1296172-33-2

Order Now

Specification Test Results

Melting point 183-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR