4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)morpholine

059823-500MG 059823-1G 059823-5G
101919-210EA 600 USD
101919-210 101919-212 101919-214
4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine ≥95%
4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MW: 204.23 g/mol
Melting Pt: 211…212 °C
MDL Number: MFCD18904308
CAS Number: 90870-83-0

Order Now

Specification Test Results

Melting point 211-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR