Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methylidene]amino}benzoate ≥95%
Supplier: Matrix Scientific
059821-500MG
059821-1G
059821-5G
101919-198EA
600
USD
101919-198
101919-200
101919-202
Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methylidene]amino}benzoate ≥95%
Methyl-3-fluoro-4-{[(2-hydroxyphenyl)methylidene]amino}benzoate
Hazard: Irritant
Formula:
C₁₅H₁₂FNO₃ Melting Pt: 113…115 °C |
MDL Number:
MFCD18904306 CAS Number: 1296225-22-3 |
Specification Test Results
Melting point | 113-115°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...