Methyl-2-fluoro-5-formylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl-2-Fluoro-5-formyl-benzoate

059820-500MG 059820-1G
101919-194EA 600 USD
101919-194 101919-196
Methyl-2-fluoro-5-formylbenzoate ≥95%
Methyl-2-fluoro-5-formylbenzoate

Hazard: Irritant

Formula: C₉H₇FO₃
MW: 182.15 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD18904319
CAS Number: 165803-94-1

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR