Methyl-4-[(E)-[(2-chloropyridin-3-yl)methylidene]amino]-3-fluorobenzoate ≥95%

Supplier: Matrix Scientific

059819-500MG 059819-1G
101919-190EA 803.08 USD
101919-190 101919-192
Methyl-4-[(E)-[(2-chloropyridin-3-yl)methylidene]amino]-3-fluorobenzoate ≥95%
Methyl-4-[(E)-[(2-chloropyridin-3-yl)methylidene]amino]-3-fluorobenzoate

Hazard: Irritant

Formula: C₁₄H₁₀ClFN₂O₂
MW: 292.7 g/mol
Melting Pt: 132…133 °C
MDL Number: MFCD18904318
CAS Number: 1296225-23-4

Order Now

Specification Test Results

Melting point 132-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR