Methyl-4-{[(5-chloro-2-hydroxyphenyl)methylidene]amino}-3-fluorobenzoate ≥95%

Supplier: Matrix Scientific

059818-500MG 059818-1G 059818-5G
101919-184EA 600 USD
101919-184 101919-186 101919-188
Methyl-4-{[(5-chloro-2-hydroxyphenyl)methylidene]amino}-3-fluorobenzoate ≥95%
Methyl-4-{[(5-chloro-2-hydroxyphenyl)methylidene]amino}-3-fluorobenzoate

Hazard: Irritant

Formula: C₁₅H₁₁ClFNO₃
Melting Pt: 172…174 °C
MDL Number: MFCD18904310
CAS Number: 1296225-20-1

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR