Methyl-4-{[(5-bromo-2-hydroxyphenyl)methylidene]amino}-3-fluorobenzoate ≥95%

Supplier: Matrix Scientific

059817-500MG 059817-1G 059817-5G
101919-178EA 600 USD
101919-178 101919-180 101919-182
Methyl-4-{[(5-bromo-2-hydroxyphenyl)methylidene]amino}-3-fluorobenzoate ≥95%
Methyl-4-{[(5-bromo-2-hydroxyphenyl)methylidene]amino}-3-fluorobenzoate

Hazard: Irritant

Formula: C₁₅H₁₁BrFNO₃
Melting Pt: 185…187 °C
MDL Number: MFCD18904307
CAS Number: 1296225-21-2

Order Now

Specification Test Results

Melting point 185-187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR