Methyl-6-{bis[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

059816-500MG 059816-1G 059816-5G
101919-172EA 403.08 USD
101919-172 101919-174 101919-176
Methyl-6-{bis[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate ≥95%
Methyl-6-{bis[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₆
MW: 352.39 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD18904309
CAS Number: 666721-08-0

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR