2-{[5-(Methoxycarbonyl)-2-nitrophenyl]amino}acetic acid ≥95%

Supplier: Matrix Scientific

059815-500MG 059815-1G
101919-168EA 600 USD
101919-168 101919-170
2-{[5-(Methoxycarbonyl)-2-nitrophenyl]amino}acetic acid ≥95%
2-{[5-(Methoxycarbonyl)-2-nitrophenyl]amino}acetic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₆
Melting Pt: 179…181 °C
MDL Number: MFCD18904317
CAS Number: 1296172-34-3

Order Now

Specification Test Results

Melting point 179-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR