2-Fluoro-5-formylbenzoic acid ≥95%

Supplier: Matrix Scientific

059811-500MG 059811-1G 059811-5G
101919-150EA 255.38 USD
101919-150 101919-152 101919-154
2-Fluoro-5-formylbenzoic acid ≥95%
2-Fluoro-5-formylbenzoic acid

Hazard: Irritant

Formula: C₈H₅FO₃
Melting Pt: 156…158 °C
MDL Number: MFCD04972875
CAS Number: 550363-85-4

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR