Ethyl-4-iodo-1H-pyrrole-2-carboxylate ≥95%
Supplier: Matrix Scientific
059810-500MG
059810-1G
059810-5G
101919-144EA
600
USD
101919-144
101919-146
101919-148
Ethyl-4-iodo-1H-pyrrole-2-carboxylate ≥95%
Ethyl-4-iodo-1H-pyrrole-2-carboxylate
Hazard: Irritant
Formula:
C₇H₈INO₂ Melting Pt: 63…64 °C |
MDL Number:
MFCD18904321 CAS Number: 433267-56-2 |
Specification Test Results
Melting point | 63-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...