2-(Ethoxycarbonyl)-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Ethoxycarbonyl)thiazole-4-carboxylic acid

059808-500MG 059808-1G
101919-136EA 200 USD
101919-136 101919-138
2-(Ethoxycarbonyl)-1,3-thiazole-4-carboxylic acid ≥95%
2-(Ethoxycarbonyl)-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₇NO₄S
MW: 201.2 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD11110995
CAS Number: 911466-96-1

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR