2,3-Dichloro-5-(chloromethyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Chloromethyl)-2,3-dichloropyridine

059804-500MG 059804-1G
101919-118EA 523.08 USD
101919-118 101919-120
2,3-Dichloro-5-(chloromethyl)pyridine ≥95%
2,3-Dichloro-5-(chloromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
Melting Pt: 36…38 °C
MDL Number: MFCD00125365
CAS Number: 54127-31-0

Order Now

Specification Test Results

Melting point 36-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR