(5-Chloro-6-methoxypyridin-3-yl)methanol ≥95%

Supplier: Matrix Scientific

059802-500MG 059802-1G
101919-110EA 669.23 USD
101919-110 101919-112
(5-Chloro-6-methoxypyridin-3-yl)methanol ≥95%
(5-Chloro-6-methoxypyridin-3-yl)methanol

Hazard: Irritant

Formula: C₇H₈ClNO₂
Melting Pt: 36…38 °C
MDL Number: MFCD04972396
CAS Number: 132865-53-3

Order Now

Specification Test Results

Melting point 36-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR