Benzyl-4-(2-oxo-1,3-diazinan-1-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

059798-500MG 059798-1G
101919-090EA 600 USD
101919-090 101919-092
Benzyl-4-(2-oxo-1,3-diazinan-1-yl)piperidine-1-carboxylate ≥95%
Benzyl-4-(2-oxo-1,3-diazinan-1-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₃N₃O₃
MW: 317.39 g/mol
Melting Pt: 194…196 °C
MDL Number: MFCD18904312
CAS Number: 164519-21-5

Order Now

Specification Test Results

Melting point 194-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR