Benzyl-4-[(3-aminopropyl)amino]piperidine-1-carboxylate dihydrochloride ≥95%

Supplier: Matrix Scientific

059797-500MG 059797-1G
101919-086EA 403.08 USD
101919-086 101919-088
Benzyl-4-[(3-aminopropyl)amino]piperidine-1-carboxylate dihydrochloride ≥95%
Benzyl-4-[(3-aminopropyl)amino]piperidine-1-carboxylate dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₇Cl₂N₃O₂
MW: 364.32 g/mol
Melting Pt: 208…210 °C
MDL Number: MFCD18904304
CAS Number: 936129-66-7

Order Now

Specification Test Results

Melting point 208-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR