Benzyl-4-[(2-aminoethyl)amino]piperidine-1-carboxylate dihydrochloride ≥95%

Supplier: Matrix Scientific

059796-500MG 059796-1G
101919-082EA 403.08 USD
101919-082 101919-084
Benzyl-4-[(2-aminoethyl)amino]piperidine-1-carboxylate dihydrochloride ≥95%
Benzyl-4-[(2-aminoethyl)amino]piperidine-1-carboxylate dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₅Cl₂N₃O₂
MW: 350.29 g/mol
Melting Pt: 212…214 °C
MDL Number: MFCD18904303
CAS Number: 1224698-21-8

Order Now

Specification Test Results

Melting point 212-214°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR