tert-Butyl-4-{[5-(methoxycarbonyl)-2-nitrophenyl]amino}piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

059666-500MG 059666-1G
101918-836EA 600 USD
101918-836 101918-838
tert-Butyl-4-{[5-(methoxycarbonyl)-2-nitrophenyl]amino}piperidine-1-carboxylate ≥95%
tert-Butyl-4-{[5-(methoxycarbonyl)-2-nitrophenyl]amino}piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₅N₃O₆
MW: 379.42 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD18803536
CAS Number: 1280787-21-4

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR