N-Phenyl-2-(trifluoromethyl)pyridin-4-amine ≥95%

Supplier: Matrix Scientific

059665-500MG 059665-1G 059665-5G
101918-830EA 803.08 USD
101918-830 101918-832 101918-834
N-Phenyl-2-(trifluoromethyl)pyridin-4-amine ≥95%
N-Phenyl-2-(trifluoromethyl)pyridin-4-amine

Hazard: Irritant

Formula: C₁₂H₉F₃N₂
Melting Pt: 135…137 °C
MDL Number: MFCD13190612
CAS Number: 234112-18-6

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR