Methyl-3-[(4-methylphenyl)sulfanyl]-4-nitrobenzoate ≥95%
Supplier: Matrix Scientific
059663-500MG
059663-1G
059663-5G
101918-820EA
600
USD
101918-820
101918-822
101918-824
Methyl-3-[(4-methylphenyl)sulfanyl]-4-nitrobenzoate ≥95%
Methyl-3-[(4-methylphenyl)sulfanyl]-4-nitrobenzoate
Hazard: Irritant
Formula:
C₁₅H₁₃NO₄S Melting Pt: 118…119 °C |
MDL Number:
MFCD18803534 CAS Number: 1280787-23-6 |
Specification Test Results
Melting point | 118-119°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...