Methyl-3-amino-4-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-nitrobenzoate

059660-500MG 059660-1G
101918-808EA 156.92 USD
101918-808 101918-810
Methyl-3-amino-4-nitrobenzoate ≥95%
Methyl-3-amino-4-nitrobenzoate

Hazard: Irritant

Formula: C₈H₈N₂O₄
Melting Pt: 201…203 °C
MDL Number: MFCD00017008
CAS Number: 99512-09-1

Order Now

Specification Test Results

Melting point 201-203°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR