Methyl-4-amino-3-fluoro-5-iodobenzoate ≥95%

Supplier: Matrix Scientific

059659-500MG 059659-1G 059659-5G
101918-802EA 707.69 USD
101918-802 101918-804 101918-806
Methyl-4-amino-3-fluoro-5-iodobenzoate ≥95%
Methyl-4-amino-3-fluoro-5-iodobenzoate

Hazard: Irritant

Formula: C₈H₇FINO₂
Melting Pt: 115…117 °C
MDL Number: MFCD13176511
CAS Number: 1123172-01-9

Order Now

Specification Test Results

Melting point 115-117°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR