Methyl-4-amino-3-bromo-5-fluorobenzoate ≥95%
Supplier: Matrix Scientific
059658-500MG
059658-1G
059658-5G
101918-796EA
203.08
USD
101918-796
101918-798
101918-800
Methyl-4-amino-3-bromo-5-fluorobenzoate ≥95%
Methyl-4-amino-3-bromo-5-fluorobenzoate
Hazard: Irritant
Formula:
C₈H₇BrFNO₂ Melting Pt: 101…102 °C |
MDL Number:
MFCD11846084 CAS Number: 1123171-91-4 |
Specification Test Results
Melting point | 101-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...