4-(4-Methoxyphenyl)-N-methyl-1,3-thiazol-2-aminehydrobromide ≥95%

Supplier: Matrix Scientific

059657-500MG 059657-1G
101918-792EA 193.85 USD
101918-792 101918-794
4-(4-Methoxyphenyl)-N-methyl-1,3-thiazol-2-aminehydrobromide ≥95%
4-(4-Methoxyphenyl)-N-methyl-1,3-thiazol-2-aminehydrobromide

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂OS
Melting Pt: 194…196 °C
MDL Number: MFCD18803531
CAS Number: 1280787-26-9

Order Now

Specification Test Results

Melting point 194-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR