2-{[5-(Methoxycarbonyl)-2-nitrophenyl]amino}propanoic acid ≥95%

Supplier: Matrix Scientific

059656-500MG 059656-1G
101918-788EA 600 USD
101918-788 101918-790
2-{[5-(Methoxycarbonyl)-2-nitrophenyl]amino}propanoic acid ≥95%
2-{[5-(Methoxycarbonyl)-2-nitrophenyl]amino}propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₆
Melting Pt: 148…150 °C
MDL Number: MFCD18803530
CAS Number: 1452523-40-8

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR