3-Iodo-5-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

059655-500MG 059655-1G 059655-5G
101918-782EA 152.31 USD
101918-782 101918-784 101918-786
3-Iodo-5-nitrobenzoic acid ≥95%
3-Iodo-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄INO₄
Melting Pt: 165…166 °C
MDL Number: MFCD00068827
CAS Number: 6313-17-3

Order Now

Specification Test Results

Melting point 165-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR