4-Fluoro-3-(4-methoxyphenyl)-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

059654-500MG 059654-1G
101918-778EA 298.46 USD
101918-778 101918-780
4-Fluoro-3-(4-methoxyphenyl)-1H-pyrazol-5-amine ≥95%
4-Fluoro-3-(4-methoxyphenyl)-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₀FN₃O
Melting Pt: 123…125 °C
MDL Number: MFCD18803529
CAS Number: 1246384-88-2

Order Now

Specification Test Results

Melting point 123-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR