2-Fluoro-3-(4-methoxyphenyl)-3-oxopropanenitrile ≥90%

Supplier: Matrix Scientific

059653-500MG 059653-1G
101918-774EA 193.85 USD
101918-774 101918-776
2-Fluoro-3-(4-methoxyphenyl)-3-oxopropanenitrile ≥90%
2-Fluoro-3-(4-methoxyphenyl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₀H₈FNO₂
Melting Pt: 79…81 °C
MDL Number: MFCD18252980
CAS Number: 1280787-24-7

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR