(4-Fluoro-1-benzothiophen-2-yl)methanol ≥95%

Supplier: Matrix Scientific

059652-500MG 059652-1G
101918-770EA 403.08 USD
101918-770 101918-772
(4-Fluoro-1-benzothiophen-2-yl)methanol ≥95%
(4-Fluoro-1-benzothiophen-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇FOS
Melting Pt: 52…54 °C
MDL Number: MFCD12032275
CAS Number: 957059-61-9

Order Now

Specification Test Results

Melting point 52-54°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR