Ethyl-4-(4-nitrophenyl)-1,3-thiazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

059651-500MG 059651-1G
101918-766EA 403.08 USD
101918-766 101918-768
Ethyl-4-(4-nitrophenyl)-1,3-thiazole-2-carboxylate ≥95%
Ethyl-4-(4-nitrophenyl)-1,3-thiazole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄S
Melting Pt: 178…180 °C
MDL Number: MFCD06797363
CAS Number: 53101-04-5

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR