Ethyl-2-(6-nitro-1,3-benzoxazol-2-yl)acetate ≥95%

Supplier: Matrix Scientific

059650-500MG 059650-1G
101918-762EA 600 USD
101918-762 101918-764
Ethyl-2-(6-nitro-1,3-benzoxazol-2-yl)acetate ≥95%
Ethyl-2-(6-nitro-1,3-benzoxazol-2-yl)acetate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₅
Melting Pt: 80…82 °C
MDL Number: MFCD00435433
CAS Number: 295345-83-4

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR