Ethyl-4-(4-chlorophenyl)-1,3-thiazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(4-chlorophenyl)thiazole-2-carboxylate

059647-500MG 059647-1G
101918-750EA 298.46 USD
101918-750 101918-752
Ethyl-4-(4-chlorophenyl)-1,3-thiazole-2-carboxylate ≥95%
Ethyl-4-(4-chlorophenyl)-1,3-thiazole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂S
MW: 267.74 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD06797356
CAS Number: 75680-91-0

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR