Ethyl-6-bromo-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

059643-500MG 059643-1G
101918-734EA 600 USD
101918-734 101918-736
Ethyl-6-bromo-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥97%
Ethyl-6-bromo-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉BrN₂O₃
Melting Pt: 264…266 °C
MDL Number: MFCD18803526
CAS Number: 894851-71-9

Order Now

Specification Test Results

Melting point 264-266°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR