Ethyl-2-(5-bromo-1H-1,3-benzodiazol-2-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(5-bromo-1H-benzo[d]imidazol-2-yl)acetate

059642-500MG 059642-1G
101918-730EA 523.08 USD
101918-730 101918-732
Ethyl-2-(5-bromo-1H-1,3-benzodiazol-2-yl)acetate ≥95%
Ethyl-2-(5-bromo-1H-1,3-benzodiazol-2-yl)acetate

Hazard: Irritant

Formula: C₁₁H₁₁BrN₂O₂
MW: 283.13 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD09953557
CAS Number: 944903-92-8

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR