Ethyl-3-amino-5-formyl-1-benzothiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

059641-500MG 059641-1G
101918-726EA 403.08 USD
101918-726 101918-728
Ethyl-3-amino-5-formyl-1-benzothiophene-2-carboxylate ≥95%
Ethyl-3-amino-5-formyl-1-benzothiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃S
Melting Pt: 158…160 °C
MDL Number: MFCD18803525
CAS Number: 1098608-18-4

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR