1,3-Diethyl-2[(2-nitrophenyl)methylidene]propanedioate ≥95%

Supplier: Matrix Scientific

059638-500MG 059638-1G 059638-5G
101918-712EA 193.85 USD
101918-712 101918-714 101918-716
1,3-Diethyl-2[(2-nitrophenyl)methylidene]propanedioate ≥95%
1,3-Diethyl-2[(2-nitrophenyl)methylidene]propanedioate

Hazard: Irritant

Formula: C₁₄H₁₅NO₆
Melting Pt: 49…51 °C
MDL Number: MFCD02176752
CAS Number: 17422-56-9

Order Now

Specification Test Results

Melting point 49-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR