2-Chloronaphthalene-1,3-dicarbaldehyde ≥95%

Supplier: Matrix Scientific

059636-500MG 059636-1G
101918-702EA 298.46 USD
101918-702 101918-704
2-Chloronaphthalene-1,3-dicarbaldehyde ≥95%
2-Chloronaphthalene-1,3-dicarbaldehyde

Hazard: Irritant

Formula: C₁₂H₇ClO₂
Melting Pt: 121…122 °C
MDL Number: MFCD18803523
CAS Number: 132554-61-1

Order Now

Specification Test Results

Melting point 121-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR