4-(4-Bromophenyl)-N-methyl-1,3-thiazol-2-aminehydrobromide ≥95%

Supplier: Matrix Scientific

059635-500MG 059635-1G
101918-698EA 193.85 USD
101918-698 101918-700
4-(4-Bromophenyl)-N-methyl-1,3-thiazol-2-aminehydrobromide ≥95%
4-(4-Bromophenyl)-N-methyl-1,3-thiazol-2-aminehydrobromide

Hazard: Irritant

Formula: C₁₀H₁₀Br₂N₂S
Melting Pt: 190…192 °C
MDL Number: MFCD18803522
CAS Number: 1280787-25-8

Order Now

Specification Test Results

Melting point 190-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR