(7-Bromo-1-benzothiophen-2-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (7-Bromobenzo[b]thiophen-2-yl)methanol, 7-Bromo-2-(hydroxymethyl)benzo[b]thiophene

059634-500MG 059634-1G
101918-694EA 315.38 USD
101918-694 101918-696
(7-Bromo-1-benzothiophen-2-yl)methanol ≥95%
(7-Bromo-1-benzothiophen-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇BrOS
MW: 243.13 g/mol
Melting Pt: 77…78 °C
MDL Number: MFCD12032282
CAS Number: 1171926-64-9

Order Now

Specification Test Results

Melting point 77-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR