2-(5-Bromo-1H-1,3-benzodiazol-2-yl)acetic acid ≥95%

Supplier: Matrix Scientific

059633-500MG 059633-1G
101918-690EA 803.08 USD
101918-690 101918-692
2-(5-Bromo-1H-1,3-benzodiazol-2-yl)acetic acid ≥95%
2-(5-Bromo-1H-1,3-benzodiazol-2-yl)acetic acid

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
Melting Pt: 148…149 °C
MDL Number: MFCD18803521
CAS Number: 1248666-87-6

Order Now

Specification Test Results

Melting point 148-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR