Bis(2,4,6-trichlorophenyl) propanedioate ≥95%
Supplier: Matrix Scientific
059632-1G
059632-5G
101918-686EA
44.62
USD
101918-686
101918-688
Bis(2,4,6-trichlorophenyl) propanedioate ≥95%
Bis(2,4,6-trichlorophenyl) propanedioate
Hazard: Irritant
Formula:
C₁₅H₆Cl₆O₄ Melting Pt: 154…156 °C |
MDL Number:
MFCD00858973 CAS Number: 15781-70-1 |
Specification Test Results
Melting point | 154-156°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...