N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide ≥95%

Supplier: Matrix Scientific

059597-500MG 059597-1G
101918-606EA 193.85 USD
101918-606 101918-608
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide ≥95%
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide

Hazard: Irritant

Formula: C₆H₁₁BrN₂S
Melting Pt: 268…270 °C
MDL Number: MFCD18651784
CAS Number: 849004-10-0

Order Now

Specification Test Results

Melting point 268-270°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR