tert-Butyl-4-(2-cyano-4-formylphenyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

059596-500MG 059596-1G 059596-5G
101918-600EA 806.15 USD
101918-600 101918-602 101918-604
tert-Butyl-4-(2-cyano-4-formylphenyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(2-cyano-4-formylphenyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₁N₃O₃
MW: 315.38 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD18651777
CAS Number: 1272756-58-7

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR