tert-Butyl-4-[(2-aminophenyl)carbonyl]piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

059595-500MG 059595-1G
101918-596EA 193.85 USD
101918-596 101918-598
tert-Butyl-4-[(2-aminophenyl)carbonyl]piperazine-1-carboxylate ≥95%
tert-Butyl-4-[(2-aminophenyl)carbonyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₃
MW: 305.38 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD09953029
CAS Number: 889125-00-2

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR