2-Nitro-3-(trifluoromethyl)phenol ≥95%

Supplier: Matrix Scientific

059592-500MG 059592-1G
101918-588EA 995.38 USD
101918-588 101918-590
2-Nitro-3-(trifluoromethyl)phenol ≥95%
2-Nitro-3-(trifluoromethyl)phenol

Hazard: Irritant

Formula: C₇H₄F₃NO₃
Melting Pt: 69…71 °C
MDL Number: MFCD08282784
CAS Number: 386-72-1

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR