2-[(4-Methylphenyl)sulfanyl]-5-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

059591-500MG 059591-1G 059591-5G
101918-582EA 193.85 USD
101918-582 101918-584 101918-586
2-[(4-Methylphenyl)sulfanyl]-5-nitrobenzonitrile ≥95%
2-[(4-Methylphenyl)sulfanyl]-5-nitrobenzonitrile

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₂S
Melting Pt: 118…120 °C
MDL Number: MFCD05365803
CAS Number: 849458-07-7

Order Now

Specification Test Results

Melting point 118-120°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR