Methyl-3-(morpholin-4-yl)-4-nitrobenzoate ≥95%
Supplier: Matrix Scientific
059588-500MG
059588-1G
059588-5G
101918-568EA
403.08
USD
101918-568
101918-570
101918-572
Methyl-3-(morpholin-4-yl)-4-nitrobenzoate ≥95%
Methyl-3-(morpholin-4-yl)-4-nitrobenzoate
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₅ Melting Pt: 90…91 °C |
MDL Number:
MFCD17780001 CAS Number: 1255785-89-7 |
Specification Test Results
Melting point | 90-91°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...