5-Formyl-2-(piperidin-1-yl)benzonitrile ≥95%

Supplier: Matrix Scientific

059583-500MG 059583-1G 059583-5G
101918-546EA 193.85 USD
101918-546 101918-548 101918-550
5-Formyl-2-(piperidin-1-yl)benzonitrile ≥95%
5-Formyl-2-(piperidin-1-yl)benzonitrile

Hazard: Irritant

Formula: C₁₃H₁₄N₂O
Melting Pt: 52…54 °C
MDL Number: MFCD18651779
CAS Number: 1272756-59-8

Order Now

Specification Test Results

Melting point 52-54°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR